Skip to Content
Merck
All Photos(1)

Documents

724238

Sigma-Aldrich

Phenyl-d5-acetyl chloride

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CH2COCl
Molecular Weight:
159.62
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

liquid

bp

94-95 °C/12 mmHg (lit.)

density

1.206 g/mL at 25 °C

mass shift

M+5

storage temp.

2-8°C
2-8°C

SMILES string

[2H]c1c([2H])c([2H])c(CC(O)=O)c([2H])c1[2H]

InChI

1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)/i1D,2D,3D,4D,5D

InChI key

WLJVXDMOQOGPHL-RALIUCGRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service