Recommended Products
Assay
95%
form
solid
mp
187-192 °C
SMILES string
[K+].F[B-](F)(F)C1CCCCC1
InChI
1S/C6H11BF3.K/c8-7(9,10)6-4-2-1-3-5-6;/h6H,1-5H2;/q-1;+1
InChI key
WWDXLJUMJZXRLX-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service