Skip to Content
Merck
All Photos(1)

Documents

722588

Sigma-Aldrich

Potassium 1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole-4-trifluoroborate

Synonym(s):

Potassium 1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazol-4-yl-4-trifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11BF3KN2O
Molecular Weight:
258.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

120-130 °C

SMILES string

[K+].F[B-](F)(F)c1cnn(c1)C2CCCCO2

InChI

1S/C8H11BF3N2O.K/c10-9(11,12)7-5-13-14(6-7)8-3-1-2-4-15-8;/h5-6,8H,1-4H2;/q-1;+1

InChI key

BFCBDXMNLWLVRE-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service