Skip to Content
Merck
All Photos(1)

Key Documents

722154

Sigma-Aldrich

4-Dodecylbenzenesulfonyl azide

(soft type) (mixture)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H29N3O2S
CAS Number:
Molecular Weight:
351.51
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:

form

liquid

impurities

≤5% ethyl acetate (residual solvent)

refractive index

n20/D 1.510

density

1.051 g/mL at 25 °C

SMILES string

CCCCCCCCCCCCc1ccc(cc1)S(=O)(=O)N=[N+]=[N-]

InChI

1S/C18H29N3O2S/c1-2-3-4-5-6-7-8-9-10-11-12-17-13-15-18(16-14-17)24(22,23)21-20-19/h13-16H,2-12H2,1H3

InChI key

DYEGEQTYEIHCSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent for synthesis of:
  • Positron emission tomography ligand for imaging metabotropic glutamate receptor type 1
  • Stitching agent for introduction of aziridines into functionalized organic molecules
  • Parenteral cephalosporins

Reagent for:
  • Intermolecular metal-catalyzed carbenoid cyclopropanations
  • Solid-phase synthesis of indolecarboxylates using palladium catalyzed reactions
  • Solid-phase rhodium carbenoid N-H insertion reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service