Skip to Content
Merck
All Photos(3)

Key Documents

719781

Sigma-Aldrich

Carbon nanofibers

greener alternative

graphitized (iron-free), composed of conical platelets, D × L 100 nm × 20-200 μm

Synonym(s):

PR-25-XT-HHT, Conical carbon nanofibers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

D × L

100 nm × 20-200 μm

surface area

24 m2/g

impurities

iron-free
<100 ppm Iron content

average diameter

130 nm

pore size

0.075 cm3/g average pore volume
124 Å average pore diameter

mp

3652-3697 °C

density

1.9 g/mL at 25 °C

bulk density

0.5‑3.5 lb/cu.ft

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Carbon nanofibers are ideal materials due to its impressive material properties such as mechanical strength, thermal and electrical conductivity. Click here for more information.

Application

Carbon nanofibers (CNFs) are carbon based nanostructures that can be used for a variety of applications such as tissue engineering, lithium based batteries, supercapacitors, and solar desalinations.

Preparation Note

Produced by Floating Catalyst Vapor-Grown Method.

Legal Information

Product of Pyrograf® Products Inc.
Pyrograf is a registered trademark of Applied Sciences, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CNFs/S 1-x Se x Composites as Promising Cathode Materials for High-Energy Lithium-Sulfur Batteries
Pandey GP, et al.
MRS Advances, 4(14), 821-828 (2019)
Electrically conductive chitosan/carbon scaffolds for cardiac tissue engineering
Martins AM, et al.
Biomacromolecules, 15(2), 635-643 (2014)
Novel draw solution for forward osmosis based solar desalination
Amjad M, et al.
Applied Energy, 230(14), 220-231 (2018)
Effect of carbon nanofibers on electrode performance of symmetric supercapcitors with composite alpha-MnO2 nanorods
Rao TP, et al.
Journal of alloys and compounds, 789(14), 518-527 (2019)
Catharina Vendl et al.
The Journal of experimental biology, 218(Pt 21), 3425-3434 (2015-11-06)
Fundamental differences in methane (CH4) production between macropods (kangaroos) and ruminants have been suggested and linked to differences in the composition of the forestomach microbiome. Using six western grey kangaroos (Macropus fuliginosus) and four red kangaroos (Macropus rufus), we measured

Articles

Pyrograf®-III vapor-grown carbon nanofibers are within the class of materials termed multi-walled carbon nanotubes (MWCNTs), and are produced by the floating catalyst method.

Composite materials with micron scale reinforcements offer tailored properties for various applications.

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Related Content

Tissue engineering fabricates tissues cultures from scaffolds, living cells, and biologically active molecules by simulating the microenvironment of the body to repair or replace damaged tissue.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service