Skip to Content
Merck
All Photos(1)

Key Documents

719684

Sigma-Aldrich

Tris(triphenylsilyloxy)molybdenum nitride pyridine complex

≥95%, contains up to 10 wt. % toluene

Synonym(s):

Molybdenum nitride tris-triphenylsilyloxide pyridine complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H50MoN2O3Si3
CAS Number:
Molecular Weight:
1015.24
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

reaction suitability

core: molybdenum
reagent type: catalyst

mp

189-191 °C

SMILES string

c1ccncc1.N#[Mo](O[Si](c2ccccc2)(c3ccccc3)c4ccccc4)(O[Si](c5ccccc5)(c6ccccc6)c7ccccc7)O[Si](c8ccccc8)(c9ccccc9)c%10ccccc%10

InChI

1S/3C18H15OSi.C5H5N.Mo.N/c3*19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-4-6-5-3-1;;/h3*1-15H;1-5H;;/q3*-1;;+3;

InChI key

OCTCTAQZWWKMIW-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service