Skip to Content
Merck
All Photos(1)

Documents

719463

Sigma-Aldrich

o-Tolylboronic acid MIDA ester

97%

Synonym(s):

2-Methylphenylboronic acid MIDA ester, 6-Methyl-2-o-tolyl-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14BNO4
Molecular Weight:
247.05
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

174-178 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccccc2C

InChI

1S/C12H14BNO4/c1-9-5-3-4-6-10(9)13-17-11(15)7-14(2)8-12(16)18-13/h3-6H,7-8H2,1-2H3

InChI key

YSHPENZDVGWTJJ-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service