Skip to Content
Merck
All Photos(1)

Key Documents

717916

Sigma-Aldrich

Chloroform solution

5% in acetone-d6 (99.9 atom % D), chromium(III) acetylacetonate 0.2 %

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€242.00

€242.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 PKG
€242.00

About This Item

MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.11

€242.00


Please contact Customer Service for Availability

Request more information

form

solution

Quality Level

composition

chromium(III) acetylacetonate, 0.2%

concentration

5% in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 7 in.
5 mm × 8 in.

suitability

suitable for NMR (reference standard)

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

1H line shape

Quantity

5 mm O.D. tube contains 0.700 mL.

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

32.0 °F - closed cup

Flash Point(C)

0 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matteo Serino et al.
PloS one, 6(6), e21184-e21184 (2011-06-24)
Daily variations in lipid concentrations in both gut lumen and blood are detected by specific sensors located in the gastrointestinal tract and in specialized central areas. Deregulation of the lipid sensors could be partly involved in the dysfunction of glucose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service