Skip to Content
Merck
All Photos(1)

Documents

717363

Sigma-Aldrich

Dichloro(DPPE)digold(I)

96%

Synonym(s):

1,2-Ethanediylbis[diphenyl]phosphine gold complex, [1,2-Bis(diphenylphosphino)ethane]dichlorodigold(I)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24Au2Cl2P2
CAS Number:
Molecular Weight:
863.26
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

reaction suitability

core: gold
reaction type: Cross Couplings
reagent type: catalyst

mp

286-289 °C

SMILES string

Cl[Au].Cl[Au].C(CP(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C26H24P2.2Au.2ClH/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)21-22-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26;;;;/h1-20H,21-22H2;;;2*1H/q;2*+1;;/p-2

InChI key

VYRCUTQUNKAXBM-UHFFFAOYSA-L

Application

Catalyst for selective diboration of alkenes via base-assisted heterolytic cleavage

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service