Skip to Content
Merck
All Photos(3)

Key Documents

716960

Sigma-Aldrich

Gold microrods

dispersion (H2O)

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€692.00

€692.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€692.00

About This Item

Empirical Formula (Hill Notation):
Au
Molecular Weight:
196.97
UNSPSC Code:
12352302
NACRES:
NA.23

€692.00


Please contact Customer Service for Availability

Request a Bulk Order

form

colloidal suspension
dispersion (H2O)

Quality Level

contains

CTAB as stabilizer

concentration

50 μg/mL in H2O

diam. × L

200 nm × 1,000 nm ± 20%

diameter

200 nm

pH

3.0-5.0

density

1 g/mL at 25 °C

Mw/Mn

(<10% CV length, monodispersity)
>95 (rods)

storage temp.

2-8°C

Physical properties

Do not freeze, quality lost if frozen even once. Please keep at approximately 4 °C

Legal Information

This product was produced under the methods claimed in U.S. Pat. Nos. 8,956,440 and 8,241,922

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Single molecule electronics is the endeavour of constructing electronic circuitry with single molecules as the fundamental building block.

Sustainable energy sources with high production efficiency are crucial for meeting increasing energy demand.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service