Skip to Content
Merck
All Photos(1)

Key Documents

716669

Sigma-Aldrich

Cadmium telluride

99.9999% trace metals basis

Synonym(s):

Cadmium monotelluride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdTe
CAS Number:
Molecular Weight:
240.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9999% trace metals basis

form

crystals

bp

1130 °C

mp

1092 °C

density

6.2 g/mL at 25 °C (lit.)

SMILES string

[Cd]=[Te]

InChI

1S/Cd.Te

InChI key

RPPBZEBXAAZZJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A direct bandgap semiconductor (bandgap 1.56 eV at 300 K). CdTe is also a strong solar cell material. It is usually sandwiched with cadmium sulfide to form a p-n junction photovoltaic solar cell. Typically, CdTe cells use a n-i-p structure.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruizheng Liang et al.
Chemical communications (Cambridge, England), 49(10), 969-971 (2012-12-13)
Ordered ultrathin films based on alternate assembly of CdTe QDs and layered double hydroxide monolayers have been fabricated, which can be used as a photoluminescence temperature sensor with dual-parameter signals and high response sensitivity.
Asad Syed et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 106, 41-47 (2013-01-30)
The growing demand for semiconductor [quantum dots (Q-dots)] nanoparticles has fuelled significant research in developing strategies for their synthesis and characterization. They are extensively investigated by the chemical route; on the other hand, use of microbial sources for biosynthesis witnessed
Chong Hyun Chang et al.
Nanoscale, 5(4), 1616-1623 (2013-01-22)
A facile electrodeposition technique was utilized to deposit single-walled carbon nanotubes (SWNTs) with cadmium telluride (CdTe) with well-controlled size, density, surface morphology, and composition. By controlling the applied charge, the morphology of these hybrid nanostructures was altered from CdTe nanoparticles
Juan P Monrás et al.
PloS one, 7(11), e48657-e48657 (2012-11-28)
The vast application of fluorescent semiconductor nanoparticles (NPs) or quantum dots (QDs) has prompted the development of new, cheap and safer methods that allow generating QDs with improved biocompatibility. In this context, green or biological QDs production represents a still
S R Stürzenbaum et al.
Nature nanotechnology, 8(1), 57-60 (2012-12-25)
The synthesis of designer solid-state materials by living organisms is an emerging field in bio-nanotechnology. Key examples include the use of engineered viruses as templates for cobalt oxide (Co(3)O(4)) particles, superparamagnetic cobalt-platinum alloy nanowires and gold-cobalt oxide nanowires for photovoltaic

Articles

Operation principle and market dominance of single crystalline silicon solar cells.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service