Skip to Content
Merck
All Photos(1)

Key Documents

715190

Sigma-Aldrich

1-Amino-3-butyne

95%

Synonym(s):

3-Butyn-1-amine, 3-Butynylamine, 4-Amino-1-butyne

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N
CAS Number:
Molecular Weight:
69.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.448

bp

100-103 °C

density

0.844 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

NCCC#C

InChI

1S/C4H7N/c1-2-3-4-5/h1H,3-5H2

InChI key

XSBPYGDBXQXSCU-UHFFFAOYSA-N

Application

1-Amino-3-butyne, a bifunctional linker can be used:
  • In Buchwald–Hartwig amination reaction of 1,2,3,4-tetrahydroacridine trifluoromethanesulfonate derivative.
  • As a reagent to synthesize dialkynylamides from diacids.
  • For the post-polymerization modification of poly(2-alkyl/aryl-2-oxazoline)s (PAOx) polymer by amidation of its methyl ester side chains.
  • As a crosslinker which can bind with resin as well as nanocrystalline cellulose to facilitate the terminal alkyne group for further functionalizations.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Predictive recognition of native proteins by cucurbit [7] uril in a complex mixture.
Li W, et al.
Chemical Communications (Cambridge, England), 52(55), 8537-8540 (2016)
Buchwald-Hartwig Amination Approach for the Synthesis of Functionalized 1, 2, 3, 4-Tetrahydroacridine Derivatives.
de Sousa J, et al.
European Journal of Organic Chemistry, 2014(16), 3468-3474 (2014)
Functional poly (2-oxazoline) s by direct amidation of methyl ester side chains.
Mees MA and Hoogenboom R
Macromolecules, 48(11), 3531-3538 (2015)
A bottom-up route to a chemically end-to-end assembly of nanocellulose fibers.
Yang H and van de Ven TGM
Biomacromolecules, 17(6), 2240-2247 (2016)
Conformationally rigid cyclic tungsten bis-alkyne complexes derived from 1, 1?-dialkynylferrocenes.
Curran TP, et al.
Journal of Organometallic Chemistry, 846(16), 24-32 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service