Skip to Content
Merck
All Photos(1)

Key Documents

715018

Sigma-Aldrich

Vinylidenecyclohexane

technical grade, 90%

Synonym(s):

Cyclohexylideneethylene, Ethenylidenecyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12
CAS Number:
Molecular Weight:
108.18
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.489

density

0.806 g/mL at 25 °C

storage temp.

−20°C

SMILES string

C=C=C1CCCCC1

InChI

1S/C8H12/c1-2-8-6-4-3-5-7-8/h1,3-7H2

InChI key

HZHTYEIEGPBKKS-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

73.9 °F - closed cup

Flash Point(C)

23.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service