Skip to Content
Merck
All Photos(1)

Key Documents

714844

Sigma-Aldrich

4-(Dimethylamino)pyridine solution

0.5 M in THF

Synonym(s):

DMAP solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
Beilstein:
3539759
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

liquid

concentration

0.5 M in THF

refractive index

n20/D 1.417

density

0.884 g/mL at 25 °C

SMILES string

CN(C)c1ccncc1

InChI

1S/C7H10N2/c1-9(2)7-3-5-8-6-4-7/h3-6H,1-2H3

InChI key

VHYFNPMBLIVWCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 2 - STOT SE 3

Target Organs

Central nervous system, Nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.0 °F

Flash Point(C)

-17.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chenghu Yan et al.
Biomacromolecules, 10(8), 2013-2018 (2009-09-03)
An effective method for grafting L-lactide (LA) from unmodified cellulose by ring-opening polymerization (ROP) in homogeneous mild conditions is presented. By using 4-dimethylaminopyridine (DMAP) as an organic catalyst, cellulose-graft-poly(L-lactide) (cellulose-g-PLLA) copolymers with a molar substitution (MS(PLLA)) of PLLA in a
Hangxiang Wang et al.
Journal of the American Chemical Society, 133(31), 12220-12228 (2011-07-19)
Catalysts hold promise as tools for chemical protein modification. However, the application of catalysts or catalyst-mediated reactions to proteins has only recently begun to be addressed, mainly in in vitro systems. By radically improving the affinity-guided DMAP (4-dimethylaminopyridine) (AGD) catalysts
A M van Wijk et al.
Analytical and bioanalytical chemistry, 400(5), 1375-1385 (2011-03-30)
A generic LC-MS/MS method was developed for the analysis of potentially genotoxic alkyl halides. A broad selection of alkyl halides were derivatized using 4-dimethylaminopyridine in acetonitrile. The reaction conditions for derivatization, i.e., solvent, reaction time, temperature and concentration of alkyl
Zheng Wang et al.
Analytical chemistry, 84(11), 4915-4920 (2012-04-28)
The development of highly sensitive and selective detection techniques for the discrimination of relevant toxic benzenethiols and biologically active aliphatic thiols is of considerable importance in the fields of chemical, biological, and environmental sciences. In this article, we describe a
Capture and visualization of hydrogen sulfide by a fluorescent probe.
Chunrong Liu et al.
Angewandte Chemie (International ed. in English), 50(44), 10327-10329 (2011-09-08)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service