Skip to Content
Merck
All Photos(1)

Key Documents

713007

Sigma-Aldrich

Butyltrimethylammonium bis(trifluoromethylsulfonyl)imide

99%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€448.00

€448.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€448.00

About This Item

Empirical Formula (Hill Notation):
C9H18F6N2O4S2
CAS Number:
Molecular Weight:
396.37
Beilstein:
9971934
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€448.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98.5% (T)
99%

form

liquid

impurities

≤0.2% water

anion traces

bromide (Br-): ≤25 mg/kg
chloride (Cl-): ≤100 mg/kg
fluoride (F-): ≤10 mg/kg
nitrate (NO3-): ≤25 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤25 mg/kg

functional group

amine
fluoro

SMILES string

CCCC[N+](C)(C)C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C7H18N.C2F6NO4S2/c1-5-6-7-8(2,3)4;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-7H2,1-4H3;/q+1;-1

InChI key

XSGKJXQWZSFJEJ-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service