Skip to Content
Merck
All Photos(3)

Key Documents

709875

Sigma-Aldrich

3-Methoxythiophene-2-boronic acid pinacol ester

97%

Synonym(s):

4,4,5,5-Tetramethyl-2-(3-methoxy-2-thienyl)-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17BO3S
CAS Number:
Molecular Weight:
240.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

124-128 °C

SMILES string

COc1ccsc1B2OC(C)(C)C(C)(C)O2

InChI

1S/C11H17BO3S/c1-10(2)11(3,4)15-12(14-10)9-8(13-5)6-7-16-9/h6-7H,1-5H3

InChI key

FXKKXDGOIQJEJS-UHFFFAOYSA-N

Application

3-Methoxythiophene-2-boronic acid pinacol ester can be used:
  • As an intermediate in the synthesis of benzothiadiazole derivatives, which are used as building blocks in the preparation of organic semiconductors.
  • As a starting material in the synthesis of imidazo[1,2-a]pyridine derivatives exhibiting bright excited-state intramolecular proton transfer (ESIPT) luminescence property in solid-state.
  • As a starting material in the preparation of polycyclic aromatic triptycenes.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of imidazo [1, 2-a] pyridine derivatives with an intramolecular hydrogen-bonded seven-membered ring exhibiting bright ESIPT luminescence in the solid state
Mutai T, et al.
Organic Letters, 21(7), 2143-2146 (2019)
Synthesis of benzothiadiazole derivatives by applying C--C cross-couplings
Heiskanen JP, et al.
The Journal of Organic Chemistry, 81(4), 1535-1546 (2016)
Polycyclic aromatic triptycenes: oxygen substitution cyclization strategies
VanVeller B, et al.
Journal of the American Chemical Society, 134(17), 7282-7285 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service