Skip to Content
Merck
All Photos(1)

Documents

708569

Sigma-Aldrich

(S)-5-Benzyl-2-mesityl-6,6-dimethyl-6,8-dihydro-5H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28BF4N3O
CAS Number:
Molecular Weight:
449.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

200-206 °C

SMILES string

F[B-](F)(F)F.CC1=C(C(C)=CC(C)=C1)[N+]2=CN3C(COC(C)(C)[C@@H]3CC4=CC=CC=C4)=N2

InChI

1S/C23H28N3O.BF4/c1-16-11-17(2)22(18(3)12-16)26-15-25-20(13-19-9-7-6-8-10-19)23(4,5)27-14-21(25)24-26;2-1(3,4)5/h6-12,15,20H,13-14H2,1-5H3;/q+1;-1/t20-;/m0./s1

InChI key

PVYSZYZMWZCJCT-BDQAORGHSA-N

Application

Catalyst for:
  • Intermolecular conjugate addition of alcohols to unsaturated ketones and esters
  • Preparation of β-amino carbonyl compounds by N-heterocyclic carbene-catalyzed enantioselective Mannich reactions of a-aryloxyacetaldehydes with imines
  • Asymmetric synthesis of cyclopentenes from allyl diketones by carbene-catalyzed intramolecular aldol reaction, lactonization, and carbon dioxide expulsion
  • Preparation of esters via tandem oxidation of allylic/benzylic/propargyl alcohols or aldehyde

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service