Skip to Content
Merck
All Photos(2)

Key Documents

707597

Sigma-Aldrich

2-Benzyloxybenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€294.00

€294.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€294.00

About This Item

Empirical Formula (Hill Notation):
C14H11ClO2
CAS Number:
Molecular Weight:
246.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€294.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

63-68 °C

functional group

acyl chloride
phenyl

storage temp.

2-8°C

SMILES string

ClC(=O)c1ccccc1OCc2ccccc2

InChI

1S/C14H11ClO2/c15-14(16)12-8-4-5-9-13(12)17-10-11-6-2-1-3-7-11/h1-9H,10H2

InChI key

ITDFRSCTQXOUAC-UHFFFAOYSA-N

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service