Skip to Content
Merck
All Photos(1)

Key Documents

707325

Sigma-Aldrich

3,5-Dibromopyridine-2-carboxaldehyde

96%

Synonym(s):

3,5-Dibromopicolinaldehyde, 3,5-Dibromopyridine-2-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Br2NO
CAS Number:
Molecular Weight:
264.90
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

60-64 °C

storage temp.

2-8°C

SMILES string

Brc1cnc(C=O)c(Br)c1

InChI

1S/C6H3Br2NO/c7-4-1-5(8)6(3-10)9-2-4/h1-3H

InChI key

BKOUPZMVLRIILB-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service