Skip to Content
Merck
All Photos(1)

Key Documents

705659

Sigma-Aldrich

3,7-Dimethyluric acid-2,4,5,6-13C4-1,3,9-15N3

99 atom % 13C, 98 atom % 15N, 98% (CP)

Synonym(s):

3,7-Dimethyl-2,6,8-trihydroxypurine-13C4,15N3 (2,4,5,6-13C4, 1,3,9-15N3)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€613.00
5 MG
€2,600.00

€613.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 MG
€613.00
5 MG
€2,600.00

About This Item

Empirical Formula (Hill Notation):
13C4C3H815N3NO3
Molecular Weight:
203.11
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

€613.00


Please contact Customer Service for Availability

Request more information

isotopic purity

99 atom % 13C
98 atom % 15N

Quality Level

Assay

98% (CP)

form

powder

mp

>300 °C

mass shift

M+7

SMILES string

CN1C(=O)[15NH][13C]2=[13C]1[13C](=O)[15NH][13C](=O)[15N]2C

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service