Skip to Content
Merck
All Photos(1)

Key Documents

705578

Sigma-Aldrich

5-Acetylamino-6-amino-3-methyluracil-(ring-13C4,15N2, amino-15N)

≥98 atom %, ≥97% (CP)

Synonym(s):

N-(4-Amino-1,2,3,6-tetrahydro-1-methyl-2,6-dioxo-5-pyrimidinyl)acetimide-(ring-13C4,15N2, amino-15N), AAMU-(ring-13C4,15N2, amino-15N)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€334.00
5 MG
€1,500.00

€334.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 MG
€334.00
5 MG
€1,500.00

About This Item

Empirical Formula (Hill Notation):
13C4C3H1015N3NO3
CAS Number:
Molecular Weight:
205.13
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.24

€334.00


Please contact Customer Service for Availability

Request more information

isotopic purity

≥98 atom %

Assay

≥97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

305 °C

mass shift

M+7

storage temp.

−20°C

SMILES string

C[15N]1[13C](=O)[15NH][13C]([15NH2])=[13C](NC(C)=O)[13C]1=O

InChI

1S/C7H10N4O3/c1-3(12)9-4-5(8)10-7(14)11(2)6(4)13/h8H2,1-2H3,(H,9,12)(H,10,14)/i4+1,5+1,6+1,7+1,8+1,10+1,11+1

InChI key

POQOTWQIYYNXAT-TUCXKDRYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service