Skip to Content
Merck
All Photos(1)

Key Documents

704229

Sigma-Aldrich

Nickel oxide - Yttria-stabilized zirconia

≥99%, general applications, NiO/YSZ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
60 wt. % NiO · 40 wt. % (Y2O3ZrO2)
UNSPSC Code:
26111700
Pricing and availability is not currently available.

Assay

≥99%

form

powder

composition

NiO, 60 wt. %
Y2O3-ZrO2, 40 wt. %

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 4 - Carc. 1A Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service