Skip to Content
Merck
All Photos(3)

Key Documents

704148

Sigma-Aldrich

Carbon nanotube, single-walled

greener alternative

(6,5) chirality, ≥90% carbon basis (≥99% as carbon nanotubes), 0.78 nm average diameter, avg. no. of layers, 1

Synonym(s):

CHASM, CNT, Signis® SG65, SWCNT, SWNT, Single wall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103

description

G/D Ratio: ≥20 (Raman 633 nm)
Median length: 1 μm

Assay

≥90% carbon basis (≥99% as carbon nanotubes)

form

powder (freeze-dried)

feature

avg. no. of layers 1

manufacturer/tradename

Signis® SG65

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

surface area

≥700 m2/g

impurities

≤5 wt. % Moisture content

average diameter

0.78 nm

mp

3652-3697 °C (lit.)

density

1.7-1.9 g/cm3 at 25 °C (lit.)

bulk density

0.1 g/cm3

greener alternative category

SMILES string

[C]

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced using CHASM′s patented CoMoCAT synthesis technology, Signis® SG65 is a single-wall carbon nanotube (CNT) product uniquely enriched in semiconducting tubes, with (6,5) chirality being most abundant. Approximately 95% of the CNTs in SG65 are semiconducting with approximately 41% of those tubes being (6,5) chirality.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Carbon nanotube, single-walled (SWNT) belongs to the class of carbonaceous materials with excellent physiochemical, thermo-mechanical and electrochemical properties. This material can be used in a variety of sustainable energy applications such as solar cells, photocatalysis, thin film conductors, field effect transistors (FETs), biosensor, gas sensor, supercapacitor and nanomechanical resonators.[1][2][3][4][5][6][7]
Suitable for use in printed semiconductors, photovoltaic devices, sensors, medical research, etc.

Preparation Note

CoMoCAT Catalytic Chemical Vapor Deposition (CVD) Method

Legal Information

CHASM is a trademark of Chasm Advanced Materials
CoMoCAT is a trademark of Chasm Advanced Materials
Signis is a registered trademark of Chasm Advanced Materials

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance dependence of SWCNT/n-silicon hybrid solar cells on the charge carrier concentration in silicon substrates
Saini V, et al.
Royal Society of Chemistry Advances, 5(1), 621-627 (2015)
Myung-Soo Choi et al.
Nanomaterials (Basel, Switzerland), 10(2) (2020-02-28)
A hybrid film consisting of zinc oxide nanoparticles (ZnO NPs) and carbon nanotubes (CNTs) is formed on a glass substrate using a simple and swift spin coating process for the use in ultraviolet photodetectors (UV PDs). The incorporation of various
Charge transfer from carbon nanotubes to silicon in flexible carbon nanotube/silicon solar cells
Li X, et al.
Small, 13(48), 1702387-1702387 (2017)
Single-walled carbon nanotube biosensors using aptamers as molecular recognition elements
So H, et al.
Journal of the American Chemical Society, 127(34), 11906-11907 (2005)
Enhanced sensitivity of a gas sensor incorporating single-walled carbon nanotube-polypyrrole nanocomposites
An KH, et al.
Advanced Materials, 16(12), 1005-1009 (2004)

Articles

The CoMoCAT® method of single-walled carbon nanotube (SWNT) synthesis yields high purity SWNTs with specific chiralities and narrow distributions of tube diameters.

Questions

1–3 of 3 Questions  
  1. Are these single-walled carbon nanotubes surface modified?

    1 answer
    1. No, these single-walled carbon nanotubes do not have surface modifications.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. How are these single-walled carbon nanotubes produced?

    1 answer
    1. These single-walled carbon nanotubes are produced by CoMoCAT® Catalytic Chemical Vapor Deposition (CVD).  Processing is done to remove amorphous carbon, and HF (aq) is used to remove the silica support and some metals.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service