Skip to Content
Merck
All Photos(1)

Key Documents

703710

Sigma-Aldrich

trans-(2-Cyclohexylvinyl)boronic acid MIDA ester

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20BNO4
CAS Number:
Molecular Weight:
265.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

form

powder

mp

218-223 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)\C=C\C2CCCCC2

InChI

1S/C13H20BNO4/c1-15-9-12(16)18-14(19-13(17)10-15)8-7-11-5-3-2-4-6-11/h7-8,11H,2-6,9-10H2,1H3/b8-7+

InChI key

YMOUBCDQXLQEBP-BQYQJAHWSA-N

Application

Purity excludes solvent of crystallization, may contain up to 10 wt. % DMSO

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service