Skip to Content
Merck
All Photos(3)

Key Documents

702765

Sigma-Aldrich

Sodium tert-pentoxide solution

1.4 M in THF

Synonym(s):

tert-Amyl alcohol sodium salt, Sodium tert-amoxide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€107.00
500 ML
€396.00

€107.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€107.00
500 ML
€396.00

About This Item

Empirical Formula (Hill Notation):
C5H11NaO
CAS Number:
Molecular Weight:
110.13
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

€107.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

Quality Level

concentration

1.4 M in THF

refractive index

n20/D 1.415

density

0.904 g/mL at 25 °C

SMILES string

[Na+].CCC(C)(C)[O-]

InChI

1S/C5H11O.Na/c1-4-5(2,3)6;/h4H2,1-3H3;/q-1;+1

InChI key

CGRKYEALWSRNJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brandon K Tate et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(28), 10160-10169 (2015-06-11)
Alkoxide-bridged disilver cations react with dihydrogen to form hydride-bridged cations, releasing free alcohol. Hydrogenolysis of neutral silver fluorides affords hydride-bridged disilver cations as their bifluoride salts. These reactions proceed most efficiently when the supporting ligands are expanded N-heterocyclic carbenes (NHCs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service