Skip to Content
Merck
All Photos(2)

Key Documents

702390

Sigma-Aldrich

2-Bromo-5-methoxybenzaldehyde

97%

Synonym(s):

3-Methoxy-6-bromobenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€28.40
5 G
€110.00

€28.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€28.40
5 G
€110.00

About This Item

Empirical Formula (Hill Notation):
C8H7BrO2
CAS Number:
Molecular Weight:
215.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€28.40


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

71-76 °C

functional group

aldehyde
bromo

SMILES string

COc1ccc(Br)c(C=O)c1

InChI

1S/C8H7BrO2/c1-11-7-2-3-8(9)6(4-7)5-10/h2-5H,1H3

InChI key

XNHKTMIWQCNZST-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service