Skip to Content
Merck
All Photos(2)

Key Documents

699837

Sigma-Aldrich

(S)-(−)-α,α-Di-(2-naphthyl)-2-pyrrolidine methanol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€239.00

€239.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€239.00

About This Item

Empirical Formula (Hill Notation):
C25H23NO
CAS Number:
Molecular Weight:
353.46
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€239.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

optical activity

[α]22/D -98.0°, c = 1 in methanol

mp

138-143 °C

functional group

hydroxyl

SMILES string

OC([C@@H]1CCCN1)(c2ccc3ccccc3c2)c4ccc5ccccc5c4

InChI

1S/C25H23NO/c27-25(24-10-5-15-26-24,22-13-11-18-6-1-3-8-20(18)16-22)23-14-12-19-7-2-4-9-21(19)17-23/h1-4,6-9,11-14,16-17,24,26-27H,5,10,15H2/t24-/m0/s1

InChI key

LNUDNNFEXRHHGY-DEOSSOPVSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service