Skip to Content
Merck
All Photos(1)

Documents

699411

Sigma-Aldrich

7-Azaindole-1-15N

98 atom % 15N, 97% (CP)

Synonym(s):

1H-Pyrrolo-1-15N(2,3-b)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H615NN
Molecular Weight:
119.13
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % 15N

Assay

97% (CP)

form

solid

mp

105-107 °C (lit.) (lit.)
105-107 °C (lit.)

mass shift

M+1

SMILES string

c1cnc2[15nH]ccc2c1

InChI

1S/C7H6N2/c1-2-6-3-5-9-7(6)8-4-1/h1-5H,(H,8,9)/i9+1

InChI key

MVXVYAKCVDQRLW-QBZHADDCSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Bulk stock may be available for this product. Please contact the Stable Isotope Customer Service Group at 937-859-1808 (1-800-448-9760 for US & Canada) or email us at isosales@sial.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service