Skip to Content
Merck
All Photos(3)

Key Documents

695254

Sigma-Aldrich

Calcium borohydride

greener alternative
Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€226.00

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€226.00

About This Item

Linear Formula:
Ca(BH4)2
CAS Number:
Molecular Weight:
69.76
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

form

powder

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[BH4-].[BH4-].[Ca++]

InChI

1S/2BH4.Ca/h2*1H4;/q2*-1;+2

InChI key

MOUPUCLGEWXWHJ-UHFFFAOYSA-N

General description

Calcium borohydride may be prepared using several techniques:
  • by reacting diborane with calcium hydride/alkoxides
  • by ball milling CaCl2 and NaBH4, resulting in an exchange reaction
  • by exchange reaction between CaCl2 and NaBH4 in THF
Ca(BH4)2 decomposes partially at 360oC and completely at 500oC.Ca(BH4)2 finds use in fuel cell, a study reports the lowering of hydrogen release temperature and pressure, by using TiCl3 as dopant.[1]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Calcium borohydride is a complex alkali earth metal borohydride, which has high gravimetric and volumetric hydrogen densities. It has a theoretical hydrogen capacity of 11.6 wt% and a low-reaction enthalpy of 32 kJ/mol H2. It can be used for greener applications, which include reversible hydrogen storage.[2][3][4][5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoconfinement in activated mesoporous carbon of calcium borohydride for improved reversible hydrogen storage
Comuanescu C, et al.
Nanotechnology, 23(38), 385401-385401 (2012)
Synthesis and characterisation of a mesoporous carbon/calcium borohydride nanocomposite for hydrogen storage
Ampoumogli A, et al.
International Journal of Hydrogen Energy, 37(21), 16631-16635 (2012)
Reversible hydrogen storage in calcium borohydride Ca (BH4) 2
Kim J, et al.
Scr. Mater., 58(6), 481-483 (2008)
Significantly enhanced dehydrogenation properties of calcium borohydride combined with urea
Chu H, et al.
Dalton Transactions, 43(41), 15291-15294 (2014)
Metal Borohydrides as Hydrogen Storage Materials
Soloveichik GL
Material Matters, 2(2), 11-11 (2007)

Articles

An article about metal borohydrides as hydrogen storage materials

Mechanochemical Effect of Severe Plastic Deformations: Metal Alloys, Hydrides and Molecular Solids

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service