Skip to Content
Merck
All Photos(1)

Key Documents

693626

Sigma-Aldrich

S-N-(3,5-Di-tert-butylphenyl)-3-methyl-2-(N-formyl-N-methylamino)butanamide

96%

Synonym(s):

Sigamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34N2O2
CAS Number:
Molecular Weight:
346.51
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

optical activity

[α]22/D -170°, c = 1 in chloroform

mp

180-186 °C

functional group

amide

storage temp.

2-8°C

SMILES string

CC(C)[C@H](N(C)C=O)C(=O)Nc1cc(cc(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C21H34N2O2/c1-14(2)18(23(9)13-24)19(25)22-17-11-15(20(3,4)5)10-16(12-17)21(6,7)8/h10-14,18H,1-9H3,(H,22,25)/t18-/m0/s1

InChI key

OVKNTNIBDGTNQY-SFHVURJKSA-N

Application

Used in the synthesis of chiral 1,2-diarylsubstituted aziridines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrei V Malkov et al.
The Journal of organic chemistry, 72(4), 1315-1325 (2007-02-10)
Asymmetric reduction of ketimines 1 with trichlorosilane can be catalyzed by N-methylvaline-derived Lewis-basic formamides 3a-d with high enantioselectivity (< or =95% ee) and low catalyst loading (1-5 mol %) at room temperature in toluene. Appending a fluorous tag, as in
Andrei V Malkov et al.
Organic letters, 6(13), 2253-2256 (2004-06-18)
[reaction: see text] Asymmetric reduction of ketimines 1 with trichlorosilane can be catalyzed by a new N-methyl L-valine derived Lewis basic organocatalyst, such as 4d, with high enantioselectivity. The structure-reactivity investigation suggests hydrogen bonding and arene-arene interactions between the catalyst
Enantioselective synthesis of 1,2-diarylaziridines by the organocatalytic reductive amination of alpha-chloroketones.
Andrei V Malkov et al.
Angewandte Chemie (International ed. in English), 46(20), 3722-3724 (2007-04-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service