Skip to Content
Merck
All Photos(1)

Key Documents

693545

Sigma-Aldrich

1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium tetrafluoroborate

95%

Synonym(s):

4,5-Dihydro-1,3-bis(2,4,6-trimethylphenyl)-1H-imidazolium tetrafluoroborate(1-), 4,5-Dihydro-1,3-dimesityl-1H-imidazolium tetrafluoroborate, 4,5-Dihydro-1,3-dimesitylimidazolium tetrafluoroborate, SIMes-HBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27BF4N2
CAS Number:
Molecular Weight:
394.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reagent type: catalyst

mp

290-296 °C

SMILES string

F[B-](F)(F)F.Cc1cc(C)c(N2CC[N+](=C2)c3c(C)cc(C)cc3C)c(C)c1

InChI

1S/C21H27N2.BF4/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;2-1(3,4)5/h9-13H,7-8H2,1-6H3;/q+1;-1

InChI key

VNRDQIOMNLIVQJ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service