Skip to Content
Merck
All Photos(1)

Key Documents

69245

Sigma-Aldrich

1-Methyl-1′-tetradecyl-4,4′-bipyridinium dichloride

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H40Cl2N2
CAS Number:
Molecular Weight:
439.50
Beilstein:
5325667
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

description

cationic

Assay

≥97.0%

mol wt

439.50 g/mol

mp

295-300 °C (dec.)

SMILES string

[Cl-].[Cl-].CCCCCCCCCCCCCC[n+]1ccc(cc1)-c2cc[n+](C)cc2

InChI

1S/C25H40N2.2ClH/c1-3-4-5-6-7-8-9-10-11-12-13-14-19-27-22-17-25(18-23-27)24-15-20-26(2)21-16-24;;/h15-18,20-23H,3-14,19H2,1-2H3;2*1H/q+2;;/p-2

InChI key

QWMLZPBYZVAWPG-UHFFFAOYSA-L

Other Notes

Amphiphilic viologen used in photochemical redox reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Krieg et al.
Journal of Colloid and Interface Science, 83, 209-209 (1981)
P.-A. Brugger et al.
Journal of the American Chemical Society, 103, 320-320 (1981)
M Grätzel et al.
Proceedings of the National Academy of Sciences of the United States of America, 80(10), 3129-3132 (1983-05-01)
Multielectron storage and hydrogen generation by light is achieved in aqueous dispersions of ultrafine TiO(2) particles (120-A diameter) when the amphiphilic viologen derivative N-tetradecyl-N'-methyl-4,4'-dipyridinium dichloride (C(14)MV(2+)) is used as an electron relay. Consecutive reduction of C(14)MV(2+) to the radical ion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service