Skip to Content
Merck
All Photos(1)

Key Documents

69227

Sigma-Aldrich

2-(Methylsulfonyl)ethyl 4-nitrophenyl carbonate

≥99.0% (C)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO7S
CAS Number:
Molecular Weight:
289.26
Beilstein:
1890569
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥99.0% (C)

mp

100-103 °C

storage temp.

2-8°C

SMILES string

CS(=O)(=O)CCOC(=O)Oc1ccc(cc1)[N+]([O-])=O

Other Notes

Reagent to introduce the MSOC-group, a versatile amino protective function with: extreme acid stability, high base lability and high polarity which enhances solubility in polar solvents.; MSOC resists hydrogenation but does not poison the catalyst

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helvetica Chimica Acta, 60, 2895-2895 (1977)
[Hormone-receptor interactions. Syntheses of alpha-melanotropin and of informational sequences thereof with the aid of alcali-labile protecting groups (author's transl)].
A Eberle et al.
Helvetica chimica acta, 58(7), 2106-2129 (1975-11-05)
G I Tesser et al.
International journal of peptide and protein research, 7(4), 295-305 (1975-01-01)
A new amino protecting group, the methylsulfonylethyloxycarbonyl (Msc) group, is described which combines well with other familiar groups (benzyloxycarbonyl, t-butyloxycarbonyl) in peptide syntheses. Its main characteristics are an extreme acid stability, a high base lability and a high polarity which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service