Skip to Content
Merck
All Photos(1)

Key Documents

690945

Sigma-Aldrich

N,N-Diethylacetoacetamide

Arxada quality, 98.0-100.3 % (w/w) (T)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€122.00
25 KG
€1,300.00
210 KG
€5,080.00

€122.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€122.00
25 KG
€1,300.00
210 KG
€5,080.00

About This Item

Linear Formula:
CH3COCH2CON(C2H5)2
CAS Number:
Molecular Weight:
157.21
Beilstein:
636486
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€122.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

concentration

98.0-100.3 % (w/w) (T)

impurities

≤0.30% water
≤4.00 g/L free amine

density

0.994 g/mL at 20 °C (lit.)

SMILES string

CCN(CC)C(=O)CC(C)=O

InChI

1S/C8H15NO2/c1-4-9(5-2)8(11)6-7(3)10/h4-6H2,1-3H3

InChI key

NTMXFHGYWJIAAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Diethylacetoacetamide is a β-dicarbonyl compound that  can be used to synthesize potent calcium channel blockers such as 2,6,6-trimethyl-3-diethylaminocarbonyl-4-aryl-5-oxo-1,4,5,6,7,8-hexahydroquinoline derivatives[1] and N,N-diethyl-2,6,6-trimethyl-4-(3-nitrophenyl)-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxamide.[2] It can also undergo Michael addition in the presence of bis(2,4-pentanedionato)nickel(II) catalyst.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on 3-Diethyl-aminocarbonyl-1, 4; 5, 6, 7, 8-hexahydroquinoline Derivatives and their Calcium Channel Antagonistic Activities in vitro.
K?smetli E
Arzneimittelforschung, 54(07), 371-375 (2004)
N, N-Diethyl-2, 6, 6-trimethyl-4-(3-nitrophenyl)-5-oxo-1, 4, 5, 6, 7, 8-hexahydroquinoline-3-carboxamide.
Linden A
Acta Crystallographica Section C, Crystal Structure Communications, 58(7), 436-438 (2002)
Nickel-catalyzed Michael additions of. beta.-dicarbonyls.
Nelson JH
The Journal of Organic Chemistry, 45(7), 1246-1249 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service