Skip to Content
Merck
All Photos(1)

Key Documents

690740

Sigma-Aldrich

Cyanuric chloride

Lonza quality, ≥99.5 % (w/w)

Synonym(s):

2,4,6-Trichloro-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3Cl3N3
CAS Number:
Molecular Weight:
184.41
Beilstein:
124246
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

vapor density

6.36 (vs air)

vapor pressure

0.8 mmHg ( 62.2 °C)

form

lumps

manufacturer/tradename

Lonza Ltd

concentration

≥99.5 % (w/w)

bp

190 °C (lit.)

mp

145-147 °C (lit.)
145.0-147.0 °C

storage temp.

2-8°C

SMILES string

Clc1nc(Cl)nc(Cl)n1

InChI

1S/C3Cl3N3/c4-1-7-2(5)9-3(6)8-1

InChI key

MGNCLNQXLYJVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

>392.0 °F - closed cup

Flash Point(C)

> 200 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramin Ghorbani-Vaghei et al.
Molecular diversity, 14(2), 385-392 (2009-08-08)
A new, facile, and efficient procedure for conjugate addition of indole and pyrrole with Michael acceptors have been developed for pulverization-activation reaction catalyzed by wet cyanuric chloride (2,4,6-trichloro-[1,3,5]-triazine or TCT) through grinding under solvent-free conditions at room temperature. Also, double-conjugate
Ravi Bhushan et al.
Amino acids, 42(4), 1371-1378 (2011-01-20)
Six dichloro-s-triazine (DCT) reagents having L-Leu, D-Phg, L-Val, L-Met, L-Ala and L-Met-NH(2) as chiral auxiliaries in cyanuric chloride were introduced for enantioseparation of 13 proteinogenic amino acids. Four other DCTs and six monochloro-s-triazine (MCT) reagents having amino acid amides as
Xue Jiang et al.
Bioresource technology, 104, 473-479 (2011-11-25)
The influence of introduction of cyanuric chloride on glucose's yield (Y) in acid-catalyzed hydrolysis of microcrystalline cellulose (MCC) has been studied. The content of cyanuric chloride (C) in modified MCCs was determined by X-ray photoelectric spectroscopy. The chemical structures of
Yu-Mee Wee et al.
Cell transplantation, 17(10-11), 1257-1269 (2009-02-03)
The necessity to transplant islet tissue without the need for immunosuppressant therapy has led to the development of materials for immune modulation. Pegylation makes islets antigenically silent, protecting them from the adsorption of foreign protein and thus avoiding immune injury.
Chao Gao et al.
Chemical communications (Cambridge, England), 49(11), 1127-1129 (2013-01-04)
A series of novel triazinonide-bridged bisimidazolium pincers were easily synthesized by quaternization of functionalized N-phenylimidazoles with highly reactive cyanuric chloride under mild conditions. The pincer was proven to be a very efficient ligand for in situ Pd-catalyzed Suzuki-Miyaura reaction with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service