Skip to Content
Merck
All Photos(2)

Key Documents

688185

Sigma-Aldrich

(Triphenylphosphoranylidene)ketene

Synonym(s):

(Triphenylphosphoranylidene)ethenone, Bestmann ylide, Ketenylidene(triphenyl)phosphorane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€94.20

€94.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€94.20

About This Item

Empirical Formula (Hill Notation):
C20H15OP
CAS Number:
Molecular Weight:
302.31
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

€94.20


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

167-173 °C

functional group

phosphine

SMILES string

O=C=C=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C20H15OP/c21-16-17-22(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20/h1-15H

InChI key

MNASRBWCHRURHY-UHFFFAOYSA-N

Application

A nucleophilic two-carbon building block that participates in a variety of reactions including cycloaddition, multi-component, and domino reactions.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Syntheses, 82, 140-140 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service