Skip to Content
Merck
All Photos(2)

Key Documents

687766

Sigma-Aldrich

7-Iodoindole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€147.00

€147.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€147.00

About This Item

Empirical Formula (Hill Notation):
C8H6IN
CAS Number:
Molecular Weight:
243.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€147.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

52-56 °C

functional group

iodo

SMILES string

Ic1cccc2cc[nH]c12

InChI

1S/C8H6IN/c9-7-3-1-2-6-4-5-10-8(6)7/h1-5,10H

InChI key

VBKAPLYISDSUCE-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chaitany Jayprakash Raorane et al.
Biomolecules, 10(8) (2020-08-23)
Multi-drug resistant Acinetobacter baumannii is well-known for its rapid acclimatization in hospital environments. The ability of the bacterium to endure desiccation and starvation on dry surfaces for up to a month results in outbreaks of health care-associated infections. Previously, indole

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service