Skip to Content
Merck
All Photos(2)

Documents

687448

Sigma-Aldrich

1-Boc-4-bromopiperidine

97%

Synonym(s):

1,1-Dimethylethyl 4-bromo-1-piperidinecarboxylate, 1-tert-Butoxycarbonyl-4-bromopiperidine, 4-Bromo-1-(tert-butoxycarbonyl)piperidine, 4-Bromo-1-piperidinecarboxylic acid tert-butyl ester, 4-Bromopiperidine-1-carboxylic acid tert-butyl ester, tert-Butyl 4-bromopiperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18BrNO2
CAS Number:
Molecular Weight:
264.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4905

bp

100-118 °C/0.5 mmHg

density

1.3403 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)N1CCC(Br)CC1

InChI

1S/C10H18BrNO2/c1-10(2,3)14-9(13)12-6-4-8(11)5-7-12/h8H,4-7H2,1-3H3

InChI key

KZBWIYHDNQHMET-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service