Skip to Content
Merck
All Photos(1)

Key Documents

68645

Sigma-Aldrich

Chloro(methyl)phenylsilane

technical, ≥93.0% (GC)

Synonym(s):

Methyl-phenyl-chlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SiH(CH3)Cl
CAS Number:
Molecular Weight:
156.68
Beilstein:
2935477
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥93.0% (GC)

form

liquid

refractive index

n20/D 1.515 (lit.)

bp

113 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

SMILES string

C[SiH](Cl)c1ccccc1

InChI

1S/C7H9ClSi/c1-9(8)7-5-3-2-4-6-7/h2-6,9H,1H3

InChI key

GVPNBWPLCHOFMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service