Skip to Content
Merck
All Photos(1)

Key Documents

685836

Sigma-Aldrich

(S)-(−)-1-Phenylethanol

97%

Synonym(s):

(−)-Methyl phenyl carbinol, (S)-(−)-α-Methylbenzyl alcohol, (S)-(−)-sec-Phenylethyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€125.00
25 G
€554.00

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€125.00
25 G
€554.00

About This Item

Empirical Formula (Hill Notation):
C8H10O
CAS Number:
Molecular Weight:
122.16
Beilstein:
2039797
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

optical activity

[α]22/D -44.0°, neat

bp

88-89 °C/10 mmHg (lit.)

mp

9-11 °C (lit.)

density

1.012 g/mL at 20 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

C[C@H](O)c1ccccc1

InChI

1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3/t7-/m0/s1

InChI key

WAPNOHKVXSQRPX-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-1-Phenylethanol can be prepared from acetophenone via bioreduction in the presence of Rhizopus arrhizus as a biocatalyst.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Laboratory scale-up synthesis of chiral carbinols using Rhizopus arrhizus.
Salvi NA and Chattopadhyay S.
Tetrahedron Asymmetry, 27(4), 188-192 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service