Skip to Content
Merck
All Photos(1)

Key Documents

680095

Sigma-Aldrich

1-Boc-4-aminopiperidine-4-carboxylic acid

97%

Synonym(s):

4-Amino-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid, 4-Amino-1-Boc-piperidine-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20N2O4
CAS Number:
Molecular Weight:
244.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Assay

97%

form

solid

mp

289-294 °C

functional group

carboxylic acid

SMILES string

CC(C)(C)OC(=O)N1CCC(N)(CC1)C(O)=O

InChI

1S/C11H20N2O4/c1-10(2,3)17-9(16)13-6-4-11(12,5-7-13)8(14)15/h4-7,12H2,1-3H3,(H,14,15)

InChI key

YNHLVALLAURVJF-UHFFFAOYSA-N

Application

Involved in investigations of conformational effects on electron-transfer efficientcy in peptide foldamers

Reactant for synthesis of:
  • SIRT2 inhibitors
  • Melanin-concentrating hormone receptor 1 antagonists
  • Bradykinin hB2 receptor antagonists
  • Neurokinin-1 receptor ligands

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service