Skip to Content
Merck
All Photos(1)

Documents

677124

Sigma-Aldrich

1,3-Cyclohexane dimethanol-d4

98 atom % D, cis/trans mixture, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D4H12O2
CAS Number:
Molecular Weight:
148.24
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

bp

283 °C

density

1.048 g/mL at 25 °C (est.)

mass shift

M+4

SMILES string

[2H]C([2H])(O)C1CCCC(C1)C([2H])([2H])O

InChI

1S/C8H16O2/c9-5-7-2-1-3-8(4-7)6-10/h7-10H,1-6H2/i5D2,6D2

InChI key

LUSFFPXRDZKBMF-NZLXMSDQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

322.0 °F - closed cup

Flash Point(C)

161.1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service