Skip to Content
Merck
All Photos(1)

Key Documents

676675

Sigma-Aldrich

(+)-9-(1S, 2S-Pseudoephedrinyl)-(10R)-(trimethylsilyl)-9-borabicyclo[3.3.2]decane

Synonym(s):

N,α-Dimethyl-β-[[(10R)-10-(trimethylsilyl)-9-borabicyclo[3.3.2]dec-9-yl]oxy]-(αS,βS)-benzeneethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H38BNOSi
CAS Number:
Molecular Weight:
371.44
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

form

solid

optical activity

[α]22/D +60°, c = 4.5 in chloroform-d

mp

130-134 °C

SMILES string

CN[C@@H](C)[C@@H](OB1C2CCCC(CCC2)[C@@H]1[Si](C)(C)C)c3ccccc3

InChI

1S/C22H38BNOSi/c1-17(24-2)21(18-11-7-6-8-12-18)25-23-20-15-9-13-19(14-10-16-20)22(23)26(3,4)5/h6-8,11-12,17,19-22,24H,9-10,13-16H2,1-5H3/t17-,19-,20+,21+,22-/m0/s1

InChI key

ZOZYQBFBLCMBTA-RXZUFOQRSA-N

Application

Air-stable, crystalline pseudoephedrine borinic acid complex which when treated with an appropriate Grignard reagent provides access to homoallylic or homopropargylic alcohols and α-allenyl carbinols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eliud Hernandez et al.
Organic letters, 7(24), 5397-5400 (2005-11-18)
[reaction: see text] The asymmetric propargylboration of aldehydes at -78 degrees C in <3 h with 1 provides silylated alpha-allenyl carbinols 6 (60-87%) in high ee (94% to >98% ee). The reagents 1 are easily prepared in both enantiomeric forms
Chunqiu Lai et al.
Organic letters, 7(5), 799-802 (2005-02-25)
The asymmetric allenylboration of representative aldehydes with the stable, storable 1 is reported. Easily and efficiently prepared in either enantiomeric form from the air-stable crystalline 4 through simple Grignard procedures, 1 gives 6 cleanly. The latter is easily isolated in
Carlos H Burgos et al.
Journal of the American Chemical Society, 127(22), 8044-8049 (2005-06-02)
The remarkable versatility and selectivity of the 10-(trimethylsilyl)-9-borabicyclo[3.3.2]decanes (10-TMS-9-BBDs) in the allyl- and crotylboration of representative aldehydes are reported. The new reagents are prepared through air-stable crystalline pseudoephedrine borinic ester complexes of the 10-TMS-9-BBDs (4), which are available in 63%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service