Skip to Content
Merck
All Photos(1)

Documents

676071

Sigma-Aldrich

Hydroxyethyl acrylate-1-13C,2,3,3-d3

97% (CP), 99 atom % 13C, 98 atom % D, contains 4-methoxyphenol as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC4D3H5O3
CAS Number:
Molecular Weight:
120.13
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

Assay

97% (CP)

contains

4-methoxyphenol as stabilizer

mass shift

M+4

storage temp.

2-8°C

SMILES string

[2H]\C([2H])=C(/[2H])[13C](=O)OCCO

InChI

1S/C5H8O3/c1-2-5(7)8-4-3-6/h2,6H,1,3-4H2/i1D2,2D,5+1

InChI key

OMIGHNLMNHATMP-YJHZBZHESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service