Skip to Content
Merck
All Photos(3)

Key Documents

673862

Sigma-Aldrich

2-Nitrophenylboronic acid

≥95%

Synonym(s):

o-Nitrobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)C6H4(B(OH)2)
CAS Number:
Molecular Weight:
166.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

mp

155-160 °C

functional group

nitro

SMILES string

OB(O)c1ccccc1[N+]([O-])=O

InChI

1S/C6H6BNO4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4,9-10H

InChI key

SFUIGUOONHIVLG-UHFFFAOYSA-N

Application

Boronic acid catalyst used for:
  • Mild and selective dipolar cycloadditions of unsaturated carboxylic acids with azides
  • Transposition of allylic alcohols and Meyer-Schuster rearrangements

Reactant involved in:
  • Suzuki-Miyaura cross-coupling reactions with aryl halides or nitroarenediazonium tetrafluoroborates
  • Copper-catalyzed halogenation
  • Oxidative arylation of aminopyrazolyl disulfides
Used in a preparation of unsymmetrical 2,2′-dinitrobiphenyls via Suzuki-Miyaura cross-coupling with nitrophenyl halides.

Other Notes

May contain varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Rodríguez González et al.
The Journal of organic chemistry, 70(23), 9591-9594 (2005-11-05)
[Reaction: see text]. Mechanistic investigations and protocols for the synthesis of 2-nitrobiphenyls and 2,2'-dinitrobiphenyls are disclosed. It is revealed that obstacles appear during the transmetalation step when the phenylboronic acid is substituted with a nitro group in the 2-position, whereas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service