Skip to Content
Merck
All Photos(1)

Key Documents

669261

Sigma-Aldrich

1-Phenyl-1-pentyn-3-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O
CAS Number:
Molecular Weight:
160.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.559

density

1.336 g/mL at 25 °C

functional group

hydroxyl
phenyl

SMILES string

CCC(O)C#Cc1ccccc1

InChI

1S/C11H12O/c1-2-11(12)9-8-10-6-4-3-5-7-10/h3-7,11-12H,2H2,1H3

InChI key

QWCMSASONHVIHV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service