Skip to Content
Merck
All Photos(1)

Key Documents

669156

Sigma-Aldrich

Chlorodicarbonyl(1,2,3,4,5-pentaphenylcyclopentadienyl)ruthenium(II)

Synonym(s):

Dicarbonylchloro[(1,2,3,4,5,-η)-1,2,3,4,5-pentaphenylcyclo-2,4-pentadien-1-yl)]ruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H25ClO2Ru
CAS Number:
Molecular Weight:
638.12
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

152-167 °C

SMILES string

Cl[Ru].[C-]#[O+].[C-]#[O+].c1ccc(cc1)[C]2[C]([C]([C]([C]2c3ccccc3)c4ccccc4)c5ccccc5)c6ccccc6

InChI

1S/C35H25.2CO.ClH.Ru/c1-6-16-26(17-7-1)31-32(27-18-8-2-9-19-27)34(29-22-12-4-13-23-29)35(30-24-14-5-15-25-30)33(31)28-20-10-3-11-21-28;2*1-2;;/h1-25H;;;1H;/q;;;;+1/p-1

InChI key

NGSBLRJKXUHTOC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Synthesis of cyclopentadienyl ruthenium dicarbonyl catalyst
  • Ligand exchange
  • Formation of cyclopentadienyl ruthenium alkoxycarbonylcomplexes with coordinated C:C bonds

Catalyst for:
  • (S)-Selective dynamic kinetic resolution of secondary alcohols
  • Stereoselective synthesis of neonicorinoide pesticide derivatives
  • Divergent asymmetric synthesis of 3,5-disubstituted piperidines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service