Skip to Content
Merck
All Photos(1)

Key Documents

668818

Sigma-Aldrich

(R)-(−)-1,2,3,4-Tetrahydro-1-naphthylamine

97%

Synonym(s):

(R)-(−)-1-AminoTetralin, (R)-(−)-1-Aminotetraline, (R)-1,2,3,4-Tetrahydro-1-naphthylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H11)NH2
CAS Number:
Molecular Weight:
147.22
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.565

bp

118-120 °C/10 mbar

density

1.002 g/mL at 25 °C

SMILES string

N[C@@H]1CCCc2ccccc12

InChI

1S/C10H13N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10H,3,5,7,11H2/t10-/m1/s1

InChI key

JRZGPXSSNPTNMA-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service