Recommended Products
Assay
97%
form
solid
mp
>300 °C
SMILES string
[K+].CCC(C)[B-](F)(F)F
InChI
1S/C4H9BF3.K/c1-3-4(2)5(6,7)8;/h4H,3H2,1-2H3;/q-1;+1
InChI key
WOHBSAAEEGTTTL-UHFFFAOYSA-N
Application
Organotrifluoroborate involved in direct alkylation of heteroaryls and Suzuki cross-coupling
Organotrifluoroborates as versatile and stable boronic acid surrogates.
Organotrifluoroborates as versatile and stable boronic acid surrogates.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service