Skip to Content
Merck
All Photos(2)

Key Documents

666564

Sigma-Aldrich

N-XantPhos

97%

Synonym(s):

NiXantphos, 4,6-Bis(diphenylphosphino)-10H-phenoxazine, 4,6-Bis(diphenylphosphino)phenoxazine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€118.00
1 G
€203.00

€118.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€118.00
1 G
€203.00

About This Item

Linear Formula:
(C6H3)2NHO(P(C6H5)2)2
CAS Number:
Molecular Weight:
551.55
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€118.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

256-262 °C

functional group

phosphine

storage temp.

2-8°C

SMILES string

N1c2cccc(P(c3ccccc3)c4ccccc4)c2Oc5c1cccc5P(c6ccccc6)c7ccccc7

InChI

1S/C36H27NOP2/c1-5-15-27(16-6-1)39(28-17-7-2-8-18-28)33-25-13-23-31-35(33)38-36-32(37-31)24-14-26-34(36)40(29-19-9-3-10-20-29)30-21-11-4-12-22-30/h1-26,37H

InChI key

HSWZLYXRAOXOLL-UHFFFAOYSA-N

Application

N-XantPhos is a deprotonatable chelating aryldiphosphine ligand that can be used in:
  • The preparation of Pd-NiXantphos catalyst system for the room temperature cross-coupling reactions of unactivated aryl chlorides.[1]
  • The synthesis of cinchonine iridium(III) cyclometalated complex that exhibits luminescence and good quantum efficiency.[2]
  • N-XantPhos and N-modified counterparts are also used in the preparation of rhodium based catalysts for hydroformylation reaction.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new approach to dendritic supported NIXANTPHOS-based hydroformylation catalysts.
Ricken S, et al.
J. Mol. Catal. A: Chem., 257(1-2), 78-88 (2006)
Enhanced emission and analyte sensing by cinchonine iridium (III) cyclometalated complexes bearing bent diphosphine chelators.
Luo S X, et al.
Organometallics, 32(10), 2908-2917 (2013)
NiXantphos: A Deprotonatable Ligand for Room-Temperature Palladium-Catalyzed Cross-Couplings of Aryl Chlorides.
Zhang J, et al.
Journal of the American Chemical Society, 136(17), 6276-6287 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service